Learn More
AdipoGen 2'-Deoxy-5-formyl-cytidine (1 mg)
SDP
Supplier:  AdipoGen AGCR13531M001
Encompass_Preferred
Chemical. CAS: 137017-45-9. Formula: C10H13N3O5. MW: 255.2. Synthetic. Epigenetic base. A minor nucleotide found in DNA and produced by UV or γ-mediated oxidation of the nucleoside 5-Methyl-2'-deoxycytidine. This nucleoside derivative was found in embryonic stem cell DNA. Mutagenic and can induce 5-fC >> T transition mutation as well as the 5-fC >> A and 5-fC >> G transversion mutations during DNA synthesis, particularly in methylated CpG regions. Used in epigenetic research and important for cancer research.
Catalog No. 50-205-3920