Learn More
Medchemexpress LLC 2′-deoxy-5-methylisocytidine | 19316-88-2 | 99.6% | 241.24 | C10H15N3O4 | 10 MG
SDP
Catalog No. 5000285862
Encompass_Preferred
2′-Deoxy-5-methylisocytidine is a deoxy nucleoside analog used as a research reagent in nucleoside chemistry and nucleic acid studies. It is provided as a solid powder intended for laboratory research use only and is typically stored at -20°C to preserve stability.
  • Purine nucleoside analog suitable for biochemical and nucleic acid research.
  • High reported purity (~99.6%) supports reproducible experimental results.
  • Supplied as a powder for flexible handling and dissolution in common solvents.
  • Suitable for synthesis, analytical characterization, and method development.
  • Long-term stability when stored at -20°C under recommended conditions.
Catalog No. 50-002-85862 Supplier Medchemexpress LLC Supplier No. HY15469210MG
2′-Deoxy-5-methylisocytidine is a deoxy nucleoside analog used as a research reagent in nucleoside chemistry and nucleic acid studies. It is provided as a solid powder intended for laboratory research use only and is typically stored at -20°C to preserve stability.
  • Purine nucleoside analog suitable for biochemical and nucleic acid research.
  • High reported purity (~99.6%) supports reproducible experimental results.
  • Supplied as a powder for flexible handling and dissolution in common solvents.
  • Suitable for synthesis, analytical characterization, and method development.
  • Long-term stability when stored at -20°C under recommended conditions.