Learn More
Medchemexpress LLC 2'-Deoxy-5,N4-dimethylcytidine | 25406-44-4 | 97.5% | 255.27 | C11H17N3O4 | 10 MG
SDP
Catalog No. 5000285974
Encompass_Preferred
2'-Deoxy-5,N4-dimethylcytidine is a purine nucleoside analog provided as an off-white to light yellow solid for laboratory research. It is used as a biochemical reagent in studies of DNA synthesis and apoptosis and is supplied at high reported purity. Handle and store according to supplier recommendations for frozen storage when required.
  • High purity suitable for research applications.
  • Off-white to light yellow solid form for easy handling.
  • Multiple small package sizes available for flexibility.
  • Stable when stored at -20°C in powder form.
  • Useful as a biochemical reagent in nucleoside and antitumor studies.
Catalog No. 50-002-85974 Supplier Medchemexpress LLC Supplier No. HY15451310MG
2'-Deoxy-5,N4-dimethylcytidine is a purine nucleoside analog provided as an off-white to light yellow solid for laboratory research. It is used as a biochemical reagent in studies of DNA synthesis and apoptosis and is supplied at high reported purity. Handle and store according to supplier recommendations for frozen storage when required.
  • High purity suitable for research applications.
  • Off-white to light yellow solid form for easy handling.
  • Multiple small package sizes available for flexibility.
  • Stable when stored at -20°C in powder form.
  • Useful as a biochemical reagent in nucleoside and antitumor studies.