Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.6% | 548.56 | 1 G
SDP
Catalog No. 5000452156
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative that exhibits potent anti-yellow fever (YFV) activity. It is intended for research use only.
  • Potent anti-yellow fever (YFV) activity.
  • Noncytotoxic at concentrations up to 20 mM.
Catalog No. 50-004-52156 Supplier Medchemexpress LLC Supplier No. HYW0086621G
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative that exhibits potent anti-yellow fever (YFV) activity. It is intended for research use only.
  • Potent anti-yellow fever (YFV) activity.
  • Noncytotoxic at concentrations up to 20 mM.