Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.6% | 548.56 | 1 ML
SDP
Catalog No. 5000366843
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It is for research use only.
  • Has 100% inhibition against YFV at 20 mM.
  • Noncytotoxic at concentrations up to 20 mM.
Catalog No. 50-003-66843 Supplier Medchemexpress LLC Supplier No. HYW00866210MM/1ML
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It is for research use only.
  • Has 100% inhibition against YFV at 20 mM.
  • Noncytotoxic at concentrations up to 20 mM.