Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.6% | 548.56 | 10 G
SDP
Catalog No. 5000366592
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative that demonstrates potent anti-yellow fever (YFV) activity. It achieves 100% inhibition against YFV at 20 mM and is noncytotoxic at concentrations up to 20 mM.
  • Exhibits 100% inhibition against YFV at 20 mM.
  • Proves to be noncytotoxic at concentrations up to 20 mM.
Catalog No. 50-003-66592 Supplier Medchemexpress LLC Supplier No. HYW00866210G
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative that demonstrates potent anti-yellow fever (YFV) activity. It achieves 100% inhibition against YFV at 20 mM and is noncytotoxic at concentrations up to 20 mM.
  • Exhibits 100% inhibition against YFV at 20 mM.
  • Proves to be noncytotoxic at concentrations up to 20 mM.