Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.6% | 548.56 | 100 MG
SDP

Catalog No. 5000452724
Encompass_Preferred

2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative. It exhibits potent anti-yellow fever (YFV) activity. It achieves 100% inhibition against YFV at 20 mM and is non-cytotoxic at concentrations up to 20 mM.

  • It is a nucleoside analogue that is a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity.
  • Achieves 100% inhibition against YFV at 20 mM.
  • Non-cytotoxic at concentrations up to 20 mM.
  • Appearance: solid, off-white to light yellow color.
  • Storage: room temperature in continental US; 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-52724 Supplier Medchemexpress LLC Supplier No. HYW008662100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative. It exhibits potent anti-yellow fever (YFV) activity. It achieves 100% inhibition against YFV at 20 mM and is non-cytotoxic at concentrations up to 20 mM.

  • It is a nucleoside analogue that is a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity.
  • Achieves 100% inhibition against YFV at 20 mM.
  • Non-cytotoxic at concentrations up to 20 mM.
  • Appearance: solid, off-white to light yellow color.
  • Storage: room temperature in continental US; 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.