Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.6% | 548.56 | 5 G
SDP
Catalog No. 5000453389
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It shows 100% inhibition against YFV at 20 mM and is noncytotoxic at concentrations up to 20 mM. This product is for research use only.
  • Nucleoside analogue
  • Potent anti-yellow fever (YFV) activity
  • Shows 100% inhibition against YFV at 20 mM
  • Noncytotoxic at concentrations up to 20 mM
  • Solid appearance, off-white to light yellow in color
Catalog No. 50-004-53389 Supplier Medchemexpress LLC Supplier No. HYW0086625G
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It shows 100% inhibition against YFV at 20 mM and is noncytotoxic at concentrations up to 20 mM. This product is for research use only.
  • Nucleoside analogue
  • Potent anti-yellow fever (YFV) activity
  • Shows 100% inhibition against YFV at 20 mM
  • Noncytotoxic at concentrations up to 20 mM
  • Solid appearance, off-white to light yellow in color