Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.62% | 548.56 | 250 MG
SDP
Catalog No. 5000453275
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine, a nucleoside analogue, is a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It proves to be noncytotoxic at concentrations up to 20 mM.
  • Potent anti-yellow fever (YFV) activity
  • Noncytotoxic at concentrations up to 20 mM
  • For research use only
Catalog No. 50-004-53275 Supplier Medchemexpress LLC Supplier No. 250MGHYW008662
2'-Deoxy-5'-O-DMT-2'-fluorouridine, a nucleoside analogue, is a 5'-O-DMTr-5-FUDR derivative with potent anti-yellow fever (YFV) activity. It proves to be noncytotoxic at concentrations up to 20 mM.
  • Potent anti-yellow fever (YFV) activity
  • Noncytotoxic at concentrations up to 20 mM
  • For research use only