Learn More
Medchemexpress LLC 2'-Deoxy-5'-O-DMT-2'-fluorouridine | 146954-74-7 | 98.62% | 548.56 | 500 MG
SDP
Catalog No. 5000452649
Encompass_Preferred
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative. It demonstrates potent activity against yellow fever virus (YFV).
  • Potent anti-yellow fever (YFV) activity
  • Shows 100% inhibition against YFV at 20 mM
  • Noncytotoxic at concentrations up to 20 mM
  • For research use only
Catalog No. 50-004-52649 Supplier Medchemexpress LLC Supplier No. HYW008662500MG
2'-Deoxy-5'-O-DMT-2'-fluorouridine is a nucleoside analogue and a 5'-O-DMTr-5-FUDR derivative. It demonstrates potent activity against yellow fever virus (YFV).
  • Potent anti-yellow fever (YFV) activity
  • Shows 100% inhibition against YFV at 20 mM
  • Noncytotoxic at concentrations up to 20 mM
  • For research use only