Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp 2-Deoxy-D-galactose 5 G
SDP

Catalog No. 502230236
Encompass_Preferred

Molecular Formula: C6H12O5
Molecular Weight: 164.16
CAS Number:?1949-89-9

Catalog No. 50-223-0236 Supplier Research Products International Corp Supplier No. D550005.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula: C6H12O5
Molecular Weight: 164.16
CAS Number:?1949-89-9

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.