Learn More
Medchemexpress LLC 2-deoxy-D-glucose-13C | 201612-55-7 | MFCD00142904 | 98.0% | 165.15 g/mol | C6H12O5 (C5(13)CH12O5; one 13C) | 5 MG
SDP
Catalog No. 5000223296
Encompass_Preferred
2-Deoxy-D-glucose-13C is a stable isotope-labeled analog of 2-deoxy-D-glucose with a single 13C label. Supplied as a white to off-white solid in a 5 mg quantity, it is intended for use as an internal standard, tracer, and research reagent in metabolic and mass spectrometry applications.
  • single 13C label for isotope tracing and internal standard applications.
  • suitable for mass spectrometry and metabolic flux analysis.
  • high purity (98.0%) appropriate for analytical workflows.
  • supplied as a 5 mg vial for small-scale research studies.
  • white to off-white solid, convenient for handling in laboratory settings.
Catalog No. 50-002-23296 Supplier Medchemexpress LLC Supplier No. HY13966S25MG
2-Deoxy-D-glucose-13C is a stable isotope-labeled analog of 2-deoxy-D-glucose with a single 13C label. Supplied as a white to off-white solid in a 5 mg quantity, it is intended for use as an internal standard, tracer, and research reagent in metabolic and mass spectrometry applications.
  • single 13C label for isotope tracing and internal standard applications.
  • suitable for mass spectrometry and metabolic flux analysis.
  • high purity (98.0%) appropriate for analytical workflows.
  • supplied as a 5 mg vial for small-scale research studies.
  • white to off-white solid, convenient for handling in laboratory settings.