Learn More
Medchemexpress LLC 2-deoxy-D-glucose-13C | 201612-55-7 | MFCD08459803 | 98.0% | C5(13C)H12O5 | 10MG
SDP
Catalog No. 5000260347
Encompass_Preferred
2-Deoxy-D-glucose-13C is a 13C-labeled isotopologue of 2-deoxy-D-glucose supplied as a white to off-white solid for isotope tracing studies. It is intended for use in metabolic flux analysis, mass spectrometry, and NMR applications to investigate glucose metabolism and glycolysis-related pathways.
  • 13C-labeled isotopologue for metabolic tracing and isotope-enrichment studies.
  • Suitable for mass spectrometry and NMR-based analyses.
  • High isotopic enrichment: 99 atom % 13C (supplier reported).
  • Chemical purity approximately 98.0%.
  • Solid, white to off-white appearance for easy handling and storage.
  • Applicable to research on glycolysis inhibition and cellular metabolism.
Catalog No. 50-002-60347 Supplier Medchemexpress LLC Supplier No. HY13966S210MG
2-Deoxy-D-glucose-13C is a 13C-labeled isotopologue of 2-deoxy-D-glucose supplied as a white to off-white solid for isotope tracing studies. It is intended for use in metabolic flux analysis, mass spectrometry, and NMR applications to investigate glucose metabolism and glycolysis-related pathways.
  • 13C-labeled isotopologue for metabolic tracing and isotope-enrichment studies.
  • Suitable for mass spectrometry and NMR-based analyses.
  • High isotopic enrichment: 99 atom % 13C (supplier reported).
  • Chemical purity approximately 98.0%.
  • Solid, white to off-white appearance for easy handling and storage.
  • Applicable to research on glycolysis inhibition and cellular metabolism.