Learn More
Cayman Chemical 2-deoxy-D-Glucose-13C6, Purity: ≥95% (mixture of isomers), 10 mg
SDP
Catalog No. 502946272
Encompass_Preferred
An internal standard for the quantification of 2-deoxy-D-glucose by GC- or LC-MS
Catalog No. 50-294-6272 Supplier Cayman Chemical Supplier No. 3431210 MG
An internal standard for the quantification of 2-deoxy-D-glucose by GC- or LC-MS