Learn More
Cayman Chemical 2-deoxy-D-Glucose-13C6, Purity: ≥95% (mixture of isomers), 1 mg
SDP
Catalog No. 502950164
Encompass_Preferred
An internal standard for the quantification of 2-deoxy-D-glucose by GC- or LC-MS
Catalog No. 50-295-0164 Supplier Cayman Chemical Supplier No. 343121 MG
An internal standard for the quantification of 2-deoxy-D-glucose by GC- or LC-MS