Learn More
Medchemexpress LLC 2-Deoxy-D-glucose | 154-17-6 | 99.9% | 164.16 | 25 G
SDP
Catalog No. 5000351874
Encompass_Preferred
2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.
  • Significantly reduces the level of ATP in MCF-7 cells in a dose- and time-dependent manner.
  • Decreases phosphorylated Akt levels and increases phosphorylated AMPK and Sirt-1 levels in MCF-7 cells.
  • Increases the levels of pentose phosphate pathway (PPP) metabolites and augments the generation of NADPH by glucose-6-phosphate dehydrogenase.
  • Increases the reduced form of glutathione by 2-DG in NB4 cells.
  • Causes a 7% decrease in final weight and delays the appearance of palpable mammary carcinomas in vivo.
  • Decreased in a dose-dependent manner in rat muscle in vivo.
Catalog No. 50-003-51874 Supplier Medchemexpress LLC Supplier No. 25GHY13966
2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.
  • Significantly reduces the level of ATP in MCF-7 cells in a dose- and time-dependent manner.
  • Decreases phosphorylated Akt levels and increases phosphorylated AMPK and Sirt-1 levels in MCF-7 cells.
  • Increases the levels of pentose phosphate pathway (PPP) metabolites and augments the generation of NADPH by glucose-6-phosphate dehydrogenase.
  • Increases the reduced form of glutathione by 2-DG in NB4 cells.
  • Causes a 7% decrease in final weight and delays the appearance of palpable mammary carcinomas in vivo.
  • Decreased in a dose-dependent manner in rat muscle in vivo.