Learn More
Medchemexpress LLC HY-13966 5g , 2-Deoxy-D-glucose 2-Deoxy-D-arabino-hexose;D-Arabino-2-deoxyhexose CAS:154-17-6 Purity:98%
SDP
Catalog No. 501492183
Encompass_Preferred
Medchemexpress, HY-13966 5g 2-Deoxy-D-glucose 2-Deoxy-D-arabino-hexose;D-Arabino-2-deoxyhexose CAS:154-17-6 Formula:C6H12O5 Purity:98% 2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2183 Supplier Medchemexpress LLC Supplier No. HY139665G
Medchemexpress, HY-13966 5g 2-Deoxy-D-glucose 2-Deoxy-D-arabino-hexose;D-Arabino-2-deoxyhexose CAS:154-17-6 Formula:C6H12O5 Purity:98% 2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.