Learn More
Medchemexpress LLC HY-13966 500mg , 2-Deoxy-D-glucose CAS:154-17-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13966500MG
Encompass_Preferred
HY-13966 500mg 2-Deoxy-D-glucose CAS: 154-17-6 2-Deoxy-D-glucose is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9514