Learn More
Cayman Chemical 2-deoxy-D-Glucose-6-phosphate (sodium salt), 10 mg
SDP
Supplier:  Cayman Chemical 1714910
Encompass_Preferred
Molecular Formula C6H12O8P • Na, Purity ≥98%, Formula Weight 266.1, Synonyms: 6-(dihydrogen phosphate) 2-deoxy-D-arabino-hexose, monosodium salt, 2-Deoxyglucose-6-phosphate, 2-DG6P; A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death.
Catalog No. NC0904668