Learn More
Medchemexpress LLC 2-Deoxy-D-glucose 6-phosphate disodium | 33068-19-8 | ≥98.0% | 288.10 | 25 MG
SDP
Catalog No. 5000348155
Encompass_Preferred
2-Deoxy-D-glucose 6-phosphate disodium is produced in mammalian cells by the action of hexokinase on 2-DG. It is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.
  • Derivative of 2-Deoxy-D-glucose
  • Produced in mammalian cells by hexokinase action on 2-DG
  • Functions as a glucose analog
  • Acts as a competitive inhibitor of glucose metabolism
  • Inhibits glycolysis via hexokinase actions
Catalog No. 50-003-48155 Supplier Medchemexpress LLC Supplier No. 25MGHY139409A
2-Deoxy-D-glucose 6-phosphate disodium is produced in mammalian cells by the action of hexokinase on 2-DG. It is a glucose analog that acts as a competitive inhibitor of glucose metabolism, inhibiting glycolysis via its actions on hexokinase.
  • Derivative of 2-Deoxy-D-glucose
  • Produced in mammalian cells by hexokinase action on 2-DG
  • Functions as a glucose analog
  • Acts as a competitive inhibitor of glucose metabolism
  • Inhibits glycolysis via hexokinase actions