Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-deoxy-D-glucose 6-phosphate disodium | 33068-19-8 | MFCD00057477 | 98.0% | 288.10 g/mol | C6H11Na2O8P | 5 MG
SDP

Catalog No. 5000223224
Encompass_Preferred

2-Deoxy-D-glucose 6-phosphate disodium is the disodium salt of phosphorylated 2-deoxy-D-glucose supplied as a white to off-white solid for laboratory research. It is used to study glycolysis, glucose metabolism, and related enzymatic pathways in biochemical assays.

  • Purity 98.0%.
  • Physical form white to off-white solid.
  • Molecular weight 288.10 g/mol.
  • High water solubility (250 mg/mL); ultrasonic assistance may be needed.
  • Storage recommended at -20°C, sealed and protected from moisture.
  • Supplied in small milligram quantities suitable for biochemical assays.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-23224 Supplier Medchemexpress LLC Supplier No. HY139409A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Deoxy-D-glucose 6-phosphate disodium is the disodium salt of phosphorylated 2-deoxy-D-glucose supplied as a white to off-white solid for laboratory research. It is used to study glycolysis, glucose metabolism, and related enzymatic pathways in biochemical assays.

  • Purity 98.0%.
  • Physical form white to off-white solid.
  • Molecular weight 288.10 g/mol.
  • High water solubility (250 mg/mL); ultrasonic assistance may be needed.
  • Storage recommended at -20°C, sealed and protected from moisture.
  • Supplied in small milligram quantities suitable for biochemical assays.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.