Learn More
Medchemexpress LLC 2-deoxy-D-glucose-d | 188004-07-1 | 98.0% | 165.16 g/mol | C6H11DO5 | 5 MG
SDP
Catalog No. 5000223295
Encompass_Preferred
2-Deoxy-D-glucose-d is a deuterium-labeled analog of 2-deoxy-D-glucose used as a stable-isotope tracer and reagent for studies of glucose metabolism. It functions as a competitive glucose analog and is used in biochemical assays to probe glycolysis and hexokinase-related pathways.
  • Deuterium-labeled analog suitable for metabolic tracing.
  • Useful for studying glycolysis and hexokinase activity.
  • High purity (98.0%), suitable for analytical applications.
  • Supplied in small research-scale quantities for isotope-labeling experiments.
  • Molecular formula C6H11DO5; molecular weight 165.16 g/mol.
Catalog No. 50-002-23295 Supplier Medchemexpress LLC Supplier No. HY13966S5MG
2-Deoxy-D-glucose-d is a deuterium-labeled analog of 2-deoxy-D-glucose used as a stable-isotope tracer and reagent for studies of glucose metabolism. It functions as a competitive glucose analog and is used in biochemical assays to probe glycolysis and hexokinase-related pathways.
  • Deuterium-labeled analog suitable for metabolic tracing.
  • Useful for studying glycolysis and hexokinase activity.
  • High purity (98.0%), suitable for analytical applications.
  • Supplied in small research-scale quantities for isotope-labeling experiments.
  • Molecular formula C6H11DO5; molecular weight 165.16 g/mol.