Learn More
Medchemexpress LLC 2-Deoxy-D-glucose-d | 188004-07-1 | MFCD35061716 | 98.0% | C6H11DO5 | 10MG
SDP
Catalog No. 5000260431
Encompass_Preferred
2-Deoxy-D-glucose-d (2-DG-d1) is a deuterium-labeled stable isotope of 2-deoxy-D-glucose used as a metabolic probe and analytical standard. It acts as a glucose analog and competitive inhibitor of glycolysis, suitable for tracer studies and mass spectrometry-based metabolomics. Molecular formula C6H11DO5; molecular weight 165.16; CAS 188004-07-1.
  • Deuterium-labeled internal standard for metabolic tracing.
  • Competitive inhibitor of glucose metabolism and glycolysis.
  • Suitable for mass spectrometry and metabolomics analyses.
  • High purity (98.0%) for research applications.
  • Available in small quantities for analytical workflows.
Catalog No. 50-002-60431 Supplier Medchemexpress LLC Supplier No. HY13966S10MG
2-Deoxy-D-glucose-d (2-DG-d1) is a deuterium-labeled stable isotope of 2-deoxy-D-glucose used as a metabolic probe and analytical standard. It acts as a glucose analog and competitive inhibitor of glycolysis, suitable for tracer studies and mass spectrometry-based metabolomics. Molecular formula C6H11DO5; molecular weight 165.16; CAS 188004-07-1.
  • Deuterium-labeled internal standard for metabolic tracing.
  • Competitive inhibitor of glucose metabolism and glycolysis.
  • Suitable for mass spectrometry and metabolomics analyses.
  • High purity (98.0%) for research applications.
  • Available in small quantities for analytical workflows.