Learn More
Apexbio Technology LLC 2-Deoxy-D-ribose(Synonyms: 2-Deoxy-D-arabinose, D-2-Deoxyribose, 2-Deoxyribose, D-Deoxyribose, D-erythro-Pentose), 100mg, CAS: 533-67-5.
SDP
Catalog No. 5000060098
Encompass_Preferred
2-Deoxy-D-ribose (CAS 533-67-5) is a naturally occurring pentose analog commonly utilized in organic synthesis and biomedical research This compound exerts biological activity by triggering apoptosis through suppression of glutathione synthesis and enhancement of cellular glutathione export sensitizing cells toward oxidative stress-induced apoptosis Additionally 2-deoxy-D-ribose serves as an intermediate in the stereoselective preparation of dipyrrolyl alkanols from pyrrole derivatives in reactions catalyzed by montmorillonite KSF clay
Catalog No. 50-000-60098 Supplier Apexbio Technology LLC Supplier No. B7864100
2-Deoxy-D-ribose (CAS 533-67-5) is a naturally occurring pentose analog commonly utilized in organic synthesis and biomedical research This compound exerts biological activity by triggering apoptosis through suppression of glutathione synthesis and enhancement of cellular glutathione export sensitizing cells toward oxidative stress-induced apoptosis Additionally 2-deoxy-D-ribose serves as an intermediate in the stereoselective preparation of dipyrrolyl alkanols from pyrrole derivatives in reactions catalyzed by montmorillonite KSF clay