Learn More
Cayman Chemical 2-deoxy-D-Ribose, Purity: ≥95%, CAS: 533-67-5, 5 g
SDP
Catalog No. 502940445
Encompass_Preferred
A reducing sugar; increases levels of ROS in HL-60 human leukemia cells at 15 mM; induces tubulogenesis and migration of BAE cells at 10 µM; topical administration of 2-deoxy-D-ribose increases blood vessel formation and accelerates wound healing in a rat full-thickness cutaneous wound model
Catalog No. 50-294-0445 Supplier Cayman Chemical Supplier No. 290725 G
A reducing sugar; increases levels of ROS in HL-60 human leukemia cells at 15 mM; induces tubulogenesis and migration of BAE cells at 10 µM; topical administration of 2-deoxy-D-ribose increases blood vessel formation and accelerates wound healing in a rat full-thickness cutaneous wound model