Learn More
Bioworld 2'-Deoxy-D-Ribose, 5 g
Catalog No. 50255896
Encompass_Preferred
Reagent used in tissue culture media. Specifications:Also known as 3-Deoxy-d-erythro-pentoseUltra Pure GradeSpecific Rotation (1%, water): -56 to-58Moisture: <0.1%Iron: <0.0005%Appearance: White powderChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 89-90 °C(lit.)RTECS Number: SB7230000
Catalog No. 50-255-896 Supplier Bioworld Supplier No. 404900081
Reagent used in tissue culture media. Specifications:Also known as 3-Deoxy-d-erythro-pentoseUltra Pure GradeSpecific Rotation (1%, water): -56 to-58Moisture: <0.1%Iron: <0.0005%Appearance: White powderChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 89-90 °C(lit.)RTECS Number: SB7230000