Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylcatechol, 99%

Catalog No. AC154350025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-29-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002201
InChI Key XESZUVZBAMCAEJ-UHFFFAOYSA-N
Synonym 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
PubChem CID 7381
IUPAC Name 4-tert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Density 1.0490g/mL
Boiling Point 285.0°C
Flash Point 129°C
Assay Percent Range 0.7% max. Catechol (GC)
Packaging Plastic Bottle
Linear Formula (CH3)3CC6H3(OH)2
Quantity 2.5 kg
Beilstein 06,IV,6014
Fieser 15,54
Specific Gravity 1.049
Solubility Information Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g
Viscosity 90 mm2/s (25°C)
Formula Weight 166.22
Percent Purity 99%
Grade Technical
Chemical Name or Material 4-tert-Butylcatechol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting eff
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-653-9
RTECSNumber UX1400000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.