Learn More
Medchemexpress LLC 2′-Deoxy-GDP trisodium | 102783-74-4 | 98.8% | 493.15 | 10 MG
SDP
Catalog No. 5000454177
Encompass_Preferred
2′-Deoxy-GDP trisodium, also known as dGDP trisodium, is a guanosine monophosphate (GMP) analogue. It functions as a hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor with an IC50 of 12.5 μM. HGPRT plays a crucial role in the purine recycling pathway of protozoan parasites.
  • Guanosine monophosphate analogue.
  • Inhibits hypoxanthine phosphoribosyltransferase.
  • IC50 of 12.5 μM against HGPRT.
  • Supports research on protozoan parasites.
  • For research use only.
Catalog No. 50-004-54177 Supplier Medchemexpress LLC Supplier No. HYW01311110MG
2′-Deoxy-GDP trisodium, also known as dGDP trisodium, is a guanosine monophosphate (GMP) analogue. It functions as a hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor with an IC50 of 12.5 μM. HGPRT plays a crucial role in the purine recycling pathway of protozoan parasites.
  • Guanosine monophosphate analogue.
  • Inhibits hypoxanthine phosphoribosyltransferase.
  • IC50 of 12.5 μM against HGPRT.
  • Supports research on protozoan parasites.
  • For research use only.