Learn More
Medchemexpress LLC 2'-Deoxy-GDP trisodium | 102783-74-4 | 98.8% | 493.15 | 5 MG
SDP
Catalog No. 5000454608
Encompass_Preferred
2'-Deoxy-GDP (dGDP) trisodium is a guanosine monophosphate (GMP) analogue and a hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor with an IC50 of 12.5 μM. HGPRT is a central enzyme in the purine recycling pathway of all protozoan parasites. It is for research use only and not sold to patients.
  • HGPRT inhibitor
  • Guanosine monophosphate (GMP) analogue
Catalog No. 50-004-54608 Supplier Medchemexpress LLC Supplier No. HYW0131115MG
2'-Deoxy-GDP (dGDP) trisodium is a guanosine monophosphate (GMP) analogue and a hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor with an IC50 of 12.5 μM. HGPRT is a central enzyme in the purine recycling pathway of all protozoan parasites. It is for research use only and not sold to patients.
  • HGPRT inhibitor
  • Guanosine monophosphate (GMP) analogue