Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxy-L-guanosine | 22837-44-1 | 99.3% | 267.24 | 5 MG
SDP

Catalog No. 5000362505
Encompass_Preferred

2'-Deoxy-L-guanosine selectively inhibits D-Thymidine phosphorylation catalyzed by HSV 1 thymidine kinase. It is the L-configuration of 2'-Deoxyguanosine. In vitro, 2'-Deoxy-L-guanosine (0-500 μM) selectively inhibits D-Thymidine phosphorylation catalyzed by HSV 1 thymidine kinase.

  • Purity: 99.30%
  • Molecular weight: 267.24
  • Appearance: Solid
  • Color: White to off-white
  • Store at 4°C, protected from light and under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL
  • For research use only

Catalog No. 50-003-62505 Supplier Medchemexpress LLC Supplier No. HY1481695MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-L-guanosine selectively inhibits D-Thymidine phosphorylation catalyzed by HSV 1 thymidine kinase. It is the L-configuration of 2'-Deoxyguanosine. In vitro, 2'-Deoxy-L-guanosine (0-500 μM) selectively inhibits D-Thymidine phosphorylation catalyzed by HSV 1 thymidine kinase.

  • Purity: 99.30%
  • Molecular weight: 267.24
  • Appearance: Solid
  • Color: White to off-white
  • Store at 4°C, protected from light and under nitrogen
  • Soluble in DMSO: ≥ 100 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.