Learn More
Medchemexpress LLC 2'-Deoxy-L-guanosine | 22837-44-1 | 99.3% | 267.24 g·mol⁻¹ | C10H13N5O4 | 10 MG
SDP
Catalog No. 5000286573
Encompass_Preferred
2'-Deoxy-L-guanosine is the L-configuration of 2'-deoxyguanosine, provided as a research-grade deoxynucleoside. It selectively inhibits D-thymidine phosphorylation catalyzed by herpes simplex virus 1 thymidine kinase and is intended for biochemical and antiviral studies requiring L-configured nucleoside analogs.
  • High purity: 99.3% by HPLC.
  • Suitable for biochemical and antiviral research applications.
  • L-configured deoxynucleoside, analog of deoxyguanosine.
  • Molecular formula C10H13N5O4; molecular weight 267.24 g·mol⁻¹.
  • Available in small research quantities, e.g., 10 mg.
  • Manufacturer documentation available: data sheet, SDS, and COA.
Catalog No. 50-002-86573 Supplier Medchemexpress LLC Supplier No. HY14816910MG
2'-Deoxy-L-guanosine is the L-configuration of 2'-deoxyguanosine, provided as a research-grade deoxynucleoside. It selectively inhibits D-thymidine phosphorylation catalyzed by herpes simplex virus 1 thymidine kinase and is intended for biochemical and antiviral studies requiring L-configured nucleoside analogs.
  • High purity: 99.3% by HPLC.
  • Suitable for biochemical and antiviral research applications.
  • L-configured deoxynucleoside, analog of deoxyguanosine.
  • Molecular formula C10H13N5O4; molecular weight 267.24 g·mol⁻¹.
  • Available in small research quantities, e.g., 10 mg.
  • Manufacturer documentation available: data sheet, SDS, and COA.