Learn More
Medchemexpress LLC 2′-deoxy-β-L-uridine | 31501-19-6 | MFCD02262147 | 100.0% | 227.195 g·mol^-1 | C9H11N2O5 | 500 MG
SDP
Catalog No. 5000285218
Encompass_Preferred
2′-Deoxy-β-L-uridine is a nucleoside analogue for biochemical and antiviral research. It functions as a substrate for certain viral enzymes and, through its 5'-triphosphates, interacts with viral DNA polymerase, making it useful for enzymatic and polymerase assays. The compound is provided as a high-purity research chemical.
  • Nucleoside analogue for biochemical and antiviral research.
  • Specific substrate for viral enzymes and polymerase assay applications.
  • High reported purity: 99.96%.
  • Chemical formula C9H11N2O5; molecular weight 227.195 g·mol^-1.
  • Available in research-scale quantities (500 mg).
Catalog No. 50-002-85218 Supplier Medchemexpress LLC Supplier No. HYW353804500MG
2′-Deoxy-β-L-uridine is a nucleoside analogue for biochemical and antiviral research. It functions as a substrate for certain viral enzymes and, through its 5'-triphosphates, interacts with viral DNA polymerase, making it useful for enzymatic and polymerase assays. The compound is provided as a high-purity research chemical.
  • Nucleoside analogue for biochemical and antiviral research.
  • Specific substrate for viral enzymes and polymerase assay applications.
  • High reported purity: 99.96%.
  • Chemical formula C9H11N2O5; molecular weight 227.195 g·mol^-1.
  • Available in research-scale quantities (500 mg).