Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2′-Deoxy-β-L-uridine | 31501-19-6 | MFCD02262147 | 100.0% | 228.20 g/mol | C9H12N2O5 | 250 MG
SDP

Catalog No. 5000285166
Encompass_Preferred

2′-Deoxy-β-L-uridine is the L-enantiomer of 2′-deoxyuridine, a nucleoside analogue used in biochemical and virology research as a substrate for viral enzymes and for studies of nucleoside metabolism. Supplied as a high-purity solid for analytical and research applications.

  • High purity: 99.96%.
  • Molecular weight: 228.20 g/mol.
  • Chemical formula: C9H12N2O5.
  • CAS number: 31501-19-6.
  • Stable as a powder at -20°C for up to 3 years and at 4°C for up to 2 years.
  • In solvent, store at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Suitable as a substrate for viral enzyme assays and studies of nucleoside metabolism.

Catalog No. 50-002-85166 Supplier Medchemexpress LLC Supplier No. HYW353804250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2′-Deoxy-β-L-uridine is the L-enantiomer of 2′-deoxyuridine, a nucleoside analogue used in biochemical and virology research as a substrate for viral enzymes and for studies of nucleoside metabolism. Supplied as a high-purity solid for analytical and research applications.

  • High purity: 99.96%.
  • Molecular weight: 228.20 g/mol.
  • Chemical formula: C9H12N2O5.
  • CAS number: 31501-19-6.
  • Stable as a powder at -20°C for up to 3 years and at 4°C for up to 2 years.
  • In solvent, store at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Suitable as a substrate for viral enzyme assays and studies of nucleoside metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.