Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2′-Deoxy-N-methylguanosine | 19916-77-9 | C11H15N5O4 | 100 MG
SDP

Catalog No. 5000376682
Encompass_Preferred

2'-Deoxy-N2-methylguanosine, a DNA adduct, is classified as a purine nucleoside analog. These analogs demonstrate broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms involved include the inhibition of DNA synthesis and the induction of apoptosis.

  • Molecular weight: 281.27
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Purity: 99.82%
  • Solubility (in vitro): DMSO : 12.5 mg/mL (44.44 mM)

Catalog No. 50-003-76682 Supplier Medchemexpress LLC Supplier No. HY154406100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-N2-methylguanosine, a DNA adduct, is classified as a purine nucleoside analog. These analogs demonstrate broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms involved include the inhibition of DNA synthesis and the induction of apoptosis.

  • Molecular weight: 281.27
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Purity: 99.82%
  • Solubility (in vitro): DMSO : 12.5 mg/mL (44.44 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.