Learn More
Medchemexpress LLC 2′-Deoxy-N-methylguanosine | 19916-77-9 | C11H15N5O4 | 25 MG
SDP
Catalog No. 5000375266
Encompass_Preferred
2′-Deoxy-N-methylguanosine is a DNA adduct and a purine nucleoside analog known for its broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis.
  • DNA adduct
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Classified under nucleoside analogs
  • Relevant to cancer research including leukemia and lymphoma
Catalog No. 50-003-75266 Supplier Medchemexpress LLC Supplier No. 25MGHY154406
2′-Deoxy-N-methylguanosine is a DNA adduct and a purine nucleoside analog known for its broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis.
  • DNA adduct
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Classified under nucleoside analogs
  • Relevant to cancer research including leukemia and lymphoma