Learn More
Medchemexpress LLC 2′-Deoxy-N-methylguanosine | 19916-77-9 | C11H15N5O4 | 5 MG
SDP
Catalog No. 5000376229
Encompass_Preferred
2'-Deoxy-N2-methylguanosine, a DNA adduct, is a purine nucleoside analog. These analogs are known to have broad antitumor activity, particularly against indolent lymphoid malignancies.
  • DNA adduct
  • Purine nucleoside analog
  • Exhibits broad antitumor activity against indolent lymphoid malignancies
  • Anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis
Catalog No. 50-003-76229 Supplier Medchemexpress LLC Supplier No. HY1544065MG
2'-Deoxy-N2-methylguanosine, a DNA adduct, is a purine nucleoside analog. These analogs are known to have broad antitumor activity, particularly against indolent lymphoid malignancies.
  • DNA adduct
  • Purine nucleoside analog
  • Exhibits broad antitumor activity against indolent lymphoid malignancies
  • Anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis