Learn More
Medchemexpress LLC 2'-Deoxy-N1-methylguanosine | 5132-79-6 | C₁₁H₁₅N₅O₄ | 50 MG
SDP
Catalog No. 5000376138
Encompass_Preferred
2'-Deoxy-N1-methylguanosine is a purine nucleoside analog that exhibits broad antitumor activity, specifically targeting indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-003-76138 Supplier Medchemexpress LLC Supplier No. HY15414750MG
2'-Deoxy-N1-methylguanosine is a purine nucleoside analog that exhibits broad antitumor activity, specifically targeting indolent lymphoid malignancies. Its anticancer mechanisms include the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analog
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis