Learn More
Medchemexpress LLC 2'-Deoxy-N1-methylguanosine | 5132-79-6 | C11H15N5O4 | 25 MG
Supplier: Medchemexpress LLC 25MGHY154147
2'-Deoxy-N1-methylguanosine is a purine nucleoside analog that exhibits broad antitumor activity against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and induction of apoptosis.
- Purine nucleoside analog.
- Broad antitumor activity targeting indolent lymphoid malignancies.
- Anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis.