Learn More
TARGETMOL CHEMICALS INC 2-Deoxy-N4-methylcytidine 5MG
SDP
Catalog No. 502880267
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2’-Deoxy-N4-methylcytidine (N(3)-Methyl-2'-deoxycytidine) is a purine nucleoside analog with potential antitumor activity for the study of apoptosis. Purity 99.61%
Catalog No. 50-288-0267 Supplier TARGETMOL CHEMICALS INC Supplier No. TNU12055MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2’-Deoxy-N4-methylcytidine (N(3)-Methyl-2'-deoxycytidine) is a purine nucleoside analog with potential antitumor activity for the study of apoptosis. Purity 99.61%