Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxy-N6-methyladenosine 3',5'-bisphosphate tetrasodium salt | 1454889-37-2 | 99.6% | 576.21 g·mol⁻¹ | C11H13N5O9P2Na4·3H2O | 5 MG
SDP

Catalog No. 5000211140
Encompass_Preferred

MRS2179 tetrasodium hydrate is a competitive P2Y1 receptor antagonist used as a research reagent to inhibit P2Y1-mediated responses, including platelet aggregation. It is supplied as a white to off-white solid with high purity and is intended for laboratory research use only; store sealed and protected from moisture.

  • Competitive P2Y1 receptor antagonist (Kb ≈102 nM).
  • High purity, 99.6%, suitable for research applications.
  • Supplied as a white to off-white solid for convenient handling and weighing.
  • Chemical formula C11H13N5O9P2Na4·3H2O; molecular weight 576.21 g·mol⁻¹.
  • Storage: sealed, away from moisture; in solvent -80°C for up to 6 months, -20°C for 1 month.

Catalog No. 50-002-11140 Supplier Medchemexpress LLC Supplier No. HY101308A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MRS2179 tetrasodium hydrate is a competitive P2Y1 receptor antagonist used as a research reagent to inhibit P2Y1-mediated responses, including platelet aggregation. It is supplied as a white to off-white solid with high purity and is intended for laboratory research use only; store sealed and protected from moisture.

  • Competitive P2Y1 receptor antagonist (Kb ≈102 nM).
  • High purity, 99.6%, suitable for research applications.
  • Supplied as a white to off-white solid for convenient handling and weighing.
  • Chemical formula C11H13N5O9P2Na4·3H2O; molecular weight 576.21 g·mol⁻¹.
  • Storage: sealed, away from moisture; in solvent -80°C for up to 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.