Learn More
Cayman Chemical 2-deoxy NAD sodIum salt 600ug
SDP
Catalog No. 502918424
Encompass_Preferred
A TRPM2 agonist; increases calcium-induced currents in an inside-out patch clamp assay using HEK293 cells expressing human TRPM2 at 30 µM; is a substrate for PARPs for the ADP-ribosylation of non-histone proteins in nuclei isolated from HeLa cells and rat livers
Catalog No. 50-291-8424 Supplier Cayman Chemical Supplier No. 38386600 UG
A TRPM2 agonist; increases calcium-induced currents in an inside-out patch clamp assay using HEK293 cells expressing human TRPM2 at 30 µM; is a substrate for PARPs for the ADP-ribosylation of non-histone proteins in nuclei isolated from HeLa cells and rat livers