Learn More
Cayman Chemical 2'-Deoxyadenosine-5'-monophosphate (hydrate), Purity: ≥95%, CAS: 653-63-4, 1 g
SDP
Catalog No. 502970750
Encompass_Preferred
A derivative of AMP that has been used in the synthesis of novel photoaffinity labels for incorporation into DNA and to study adenosine-based interactions during DNA synthesis and DNA damage
Catalog No. 50-297-0750 Supplier Cayman Chemical Supplier No. 162181 G
A derivative of AMP that has been used in the synthesis of novel photoaffinity labels for incorporation into DNA and to study adenosine-based interactions during DNA synthesis and DNA damage