Learn More
Apexbio Technology LLC 2-Deoxyadenosine 958-09-8 100mg
SDP
Catalog No. 502240756
Encompass_Preferred
2-Deoxyadenosine (CAS 958-09-8) is a synthetic purine nucleoside analog that functions as a demethylated nucleoside in biological systems Structurally similar to adenosine but lacking a hydroxyl group at the 2 position it selectively pairs with deoxythymidine during DNA synthesis 2-Deoxyadenosine influences regulatory pathways associated with DNA replication and repair serving as a molecular probe to study these processes It is extensively utilized in molecular biology and genomics for investigating DNA synthesis repair mechanisms and the dynamics of nucleic acid metabolism
This item is not returnable. View return policy
Catalog No. 50-224-0756 Supplier Apexbio Technology LLC Supplier No. B7860100
2-Deoxyadenosine (CAS 958-09-8) is a synthetic purine nucleoside analog that functions as a demethylated nucleoside in biological systems Structurally similar to adenosine but lacking a hydroxyl group at the 2 position it selectively pairs with deoxythymidine during DNA synthesis 2-Deoxyadenosine influences regulatory pathways associated with DNA replication and repair serving as a molecular probe to study these processes It is extensively utilized in molecular biology and genomics for investigating DNA synthesis repair mechanisms and the dynamics of nucleic acid metabolism