Learn More
Medchemexpress LLC HY-W040329 100mg Medchemexpress, 2'-Deoxyadenosine CAS:958-09-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW040329100MG
Encompass_Preferred
Medchemexpress, HY-W040329 100mg 2'-Deoxyadenosine CAS:958-09-8 2'-Deoxyadenosine is a nucleoside adenosine derivative, pairing with deoxythymidine (T) in double-stranded DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3472