Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyadenosine-13C10 | ≥99.0% | 261.17 | 13C10H13N5O3 | 1 MG
SDP

Catalog No. 5000208731
Encompass_Preferred

2'-deoxyadenosine-13C10 is a 13C-labeled isotope standard of 2'-deoxyadenosine intended for use as an analytical reference and tracer in biochemical and mass spectrometry assays.

  • Isotope-labeled standard for analytical and tracer studies.
  • Molecular formula: 13C10H13N5O3.
  • Molecular weight: 261.17 g/mol.
  • Purity: ≥99.0%.
  • Supplied as a 1 mg vial.
  • Suitable for use in mass spectrometry and quantitative assays.

Catalog No. 50-002-08731 Supplier Medchemexpress LLC Supplier No. HYW040329S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-deoxyadenosine-13C10 is a 13C-labeled isotope standard of 2'-deoxyadenosine intended for use as an analytical reference and tracer in biochemical and mass spectrometry assays.

  • Isotope-labeled standard for analytical and tracer studies.
  • Molecular formula: 13C10H13N5O3.
  • Molecular weight: 261.17 g/mol.
  • Purity: ≥99.0%.
  • Supplied as a 1 mg vial.
  • Suitable for use in mass spectrometry and quantitative assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.