Learn More
Medchemexpress LLC 2-Deoxyadenosine 5 100mg
SDP
Catalog No. 5000206184
Encompass_Preferred
2 -Deoxyadenosine 5 -monophosphate disodium a nucleic acid AMP derivative is a deoxyribonucleotide found in DNA 2 -Deoxyadenosine 5 -monophosphate disodium can be used to study adenosine-based interactions during DNA synthesis and DNA damage[1]
Catalog No. 50-002-06184 Supplier Medchemexpress LLC Supplier No. HYW105272100MG
2 -Deoxyadenosine 5 -monophosphate disodium a nucleic acid AMP derivative is a deoxyribonucleotide found in DNA 2 -Deoxyadenosine 5 -monophosphate disodium can be used to study adenosine-based interactions during DNA synthesis and DNA damage[1]