Learn More
Medchemexpress LLC 2-Deoxyadenosine 5 250mg
SDP
Catalog No. 5000205310
Encompass_Preferred
2 -Deoxyadenosine 5 -monophosphate disodium a nucleic acid AMP derivative is a deoxyribonucleotide found in DNA 2 -Deoxyadenosine 5 -monophosphate disodium can be used to study adenosine-based interactions during DNA synthesis and DNA damage[1]
Catalog No. 50-002-05310 Supplier Medchemexpress LLC Supplier No. 250MGHYW105272
2 -Deoxyadenosine 5 -monophosphate disodium a nucleic acid AMP derivative is a deoxyribonucleotide found in DNA 2 -Deoxyadenosine 5 -monophosphate disodium can be used to study adenosine-based interactions during DNA synthesis and DNA damage[1]