Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate | 653-63-4 | 99.2% | 331.22 | C10H14N5O6P | 25 MG
SDP

Catalog No. 5000304044
Encompass_Preferred

2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxyribonucleotide used as an analytical standard and reference material for nucleic acid research, method validation, and chromatographic or mass-spectrometry assays. Supplied as a solid in amber glass packaging, it is intended for calibration, quantitation, and analytical method development.

  • High purity analytical standard suitable for HPLC and MS.
  • Purity confirmed by HPLC (99.15% reported by COA).
  • Molecular formula C10H14N5O6P and molecular weight 331.22.
  • Supplied in brown glass to protect from light and moisture.
  • Intended for calibration, method validation, and quantitative analyses.

Catalog No. 50-003-04044 Supplier Medchemexpress LLC Supplier No. 25MGHYW016009R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxyribonucleotide used as an analytical standard and reference material for nucleic acid research, method validation, and chromatographic or mass-spectrometry assays. Supplied as a solid in amber glass packaging, it is intended for calibration, quantitation, and analytical method development.

  • High purity analytical standard suitable for HPLC and MS.
  • Purity confirmed by HPLC (99.15% reported by COA).
  • Molecular formula C10H14N5O6P and molecular weight 331.22.
  • Supplied in brown glass to protect from light and moisture.
  • Intended for calibration, method validation, and quantitative analyses.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.