Learn More
Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate | 653-63-4 | 99.2% | 331.22 g/mol | C10H14N5O6P | 100 MG
SDP
Catalog No. 5000299935
Encompass_Preferred
2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxynucleotide monophosphate used as a biochemical reagent and reference standard in nucleic acid research and analytical chemistry. Supplied as a white to off-white solid, it is used in enzymatic assays, chromatography, mass spectrometry, and studies of DNA metabolism.
Catalog No. 50-002-99935 Supplier Medchemexpress LLC Supplier No. HYW016009R100MG
2'-Deoxyadenosine-5'-monophosphate (dAMP) is a deoxynucleotide monophosphate used as a biochemical reagent and reference standard in nucleic acid research and analytical chemistry. Supplied as a white to off-white solid, it is used in enzymatic assays, chromatography, mass spectrometry, and studies of DNA metabolism.