Learn More
Medchemexpress LLC 2'-deoxyadenosine-5'-monophosphate | 653-63-4 | 99.2% | 331.22 g/mol | C10H14N5O6P | 250 MG
SDP
Catalog No. 5000303730
Encompass_Preferred
2'-Deoxyadenosine-5'-monophosphate is a deoxyribonucleotide (adenosine monophosphate derivative) provided as a high-purity analytical standard for research and analytical applications, including studies of DNA synthesis and nucleoside metabolism. Supplied as a solid standard in laboratory quantities for use in biochemical assays.
Catalog No. 50-003-03730 Supplier Medchemexpress LLC Supplier No. 250MGHYW016009R
2'-Deoxyadenosine-5'-monophosphate is a deoxyribonucleotide (adenosine monophosphate derivative) provided as a high-purity analytical standard for research and analytical applications, including studies of DNA synthesis and nucleoside metabolism. Supplied as a solid standard in laboratory quantities for use in biochemical assays.